Learn More
Selleck Chemical LLC Carbenicillin disodium S3179-1g
SDP
Catalog No. 503110876
Encompass_Preferred
Carbenicillin (BRL-2064 -carboxybenzylpenicillin) is a semi-synthetic penicillin antibiotic which interferes with cell wall synthesis of gram-negative bacteria while displaying low toxicity
Catalog No. 50-311-0876 Supplier Selleck Chemical LLC Supplier No. S31791G
Carbenicillin (BRL-2064 -carboxybenzylpenicillin) is a semi-synthetic penicillin antibiotic which interferes with cell wall synthesis of gram-negative bacteria while displaying low toxicity