Learn More
Bioworld Carbenicillin Disodium Salt, 10 g
Supplier:  Bioworld 403100003
Encompass_Preferred
Carbenicillin Disodium Salt Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Analog to ampicillin. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.Potency >770 μg/mgHygroscopicUSP gradeMay cause sensitization by inhalation and skin contact.For in vitro lab use only. Not for human use or consumption.
Catalog No. 50-255-710