Learn More
Enzo Life Sciences Carbenicillin . disodium salt (1g). CAS: 4800-94-6
SDP
Supplier:  Enzo Life Sciences ALX380270G001
Encompass_Preferred
Analog to ampicillin (ALX-380-268). Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-positive and Gram-negative bacteria and Pseudomonas. Alternative name: alpha-Carboxybenzylpenicillin . 2Na. Purity: ≥92% (Assay). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9001