Learn More
Alkali Scientific Carbenicillin, Disodium Salt, 25 G
SDP
Catalog No. 5000660014
Encompass_Preferred
Carbenicillin Disodium Salt is a beta-lactam antibiotic widely used in microbiology and molecular biology for selective inhibition of bacterial growth. It acts by disrupting cell wall synthesis, leading to bacterial cell death. Carbenicillin is frequently used in cloning and transformation experiments to maintain plasmid selection in bacterial cultures. It is more stable than ampicillin and reduces the formation of satellite colonies, ensuring clearer selection results. This compound is suitable for use in laboratory media and cell culture applications where consistent antibiotic activity is required. It supports research involving genetic engineering, microbial growth control, and studies of antibiotic resistance mechanisms.
Catalog No. 50-006-60014 Supplier Alkali Scientific Supplier No. CAR003CP
Carbenicillin Disodium Salt is a beta-lactam antibiotic widely used in microbiology and molecular biology for selective inhibition of bacterial growth. It acts by disrupting cell wall synthesis, leading to bacterial cell death. Carbenicillin is frequently used in cloning and transformation experiments to maintain plasmid selection in bacterial cultures. It is more stable than ampicillin and reduces the formation of satellite colonies, ensuring clearer selection results. This compound is suitable for use in laboratory media and cell culture applications where consistent antibiotic activity is required. It supports research involving genetic engineering, microbial growth control, and studies of antibiotic resistance mechanisms.