Learn More
Carbenicillin, Disodium Salt, G-Biosciences
SCI_ED
Catalog No. S162501
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
S162501 500 g
1 options
Catalog No. S162501 Supplier G-Biosciences Supplier No. RC853
Add to Cart
Add to Cart
Carbenicillin is an inhibitor of bacterial cell wall synthesis, with a similar method of action as Ampicillin.
Highly effective against gram-negative bacteria. B-lactamase sensitive. This high grade salt is nontoxic to plant cells.
Specifications
Form Powder
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Quantity 500 g