Learn More
Ubpbio Carbenicillin DisodiumSalt, 5G
Catalog No. 501965189
Encompass_Preferred
Carbenicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria. Carbenicillin is more stable than ampicillin and results in fewer satellite colonies on selection plates.
Catalog No. 50-196-5189 Supplier Ubpbio Supplier No. P80205
Carbenicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria. Carbenicillin is more stable than ampicillin and results in fewer satellite colonies on selection plates.