Learn More
Crescent Chemical Co Inc CARBENICILLIN-NA2-SALT
SDP
Catalog No. 50247395
Encompass_Preferred
Carbenicillin-Na2-salt research grade Semisynthetic derivative of benzylpenicillin G. Inhibitor of bacterial cell wall synthesis. Active above all against gram negative bacteria, less against gram positive. Used in molecular biology for selection of resistant strains (2
Catalog No. 50-247-395 Supplier Crescent Chemical Co Inc Supplier No. SE15875.03
Carbenicillin-Na2-salt research grade Semisynthetic derivative of benzylpenicillin G. Inhibitor of bacterial cell wall synthesis. Active above all against gram negative bacteria, less against gram positive. Used in molecular biology for selection of resistant strains (2