Learn More
TOKU-E Carbenicillin ReadyMade™ Solution | Carbenicillin Disodium | CAS 4800-94-6 | 100 mg/ML in 20% ethanol | sterile-filtered | 50 x 1 ML
Catalog No. NC2068760
Encompass_Preferred
Carbenicillin ReadyMade™ Solution | Carbenicillin Disodium | CAS 4800-94-6 | 100 mg/ML in 20% ethanol | sterile-filtered | 50 x 1 ML
Catalog No. NC2068760 Supplier TOKU-E Supplier No. C28950X1ML
Carbenicillin ReadyMade™ Solution | Carbenicillin Disodium | CAS 4800-94-6 | 100 mg/ML in 20% ethanol | sterile-filtered | 50 x 1 ML