Learn More
Gold Biotechnology Inc Carbenicillin Solution
SDP
Catalog No. 5000562176
Encompass_Preferred
Carbenicillin is a member of the penicillin -lactam antibiotics However unlike most -lactams carbenicillin disodium is limited to primarily gram-negative bacteria including Pseudomonas aeruginosa and common enteric species It can be used in selection protocols and is often preferred over ampicillin due to its increased stability This allows for less contamination of unwanted bacterial colonies In addition carbenicillin has been used to regulate bacterial growth in plants yielding low regeneration frequencies It can be used in genetic transformation applications to select for AmpR transformed cells
Catalog No. 50-005-62176 Supplier Gold Biotechnology Inc Supplier No. C103SL10
Carbenicillin is a member of the penicillin -lactam antibiotics However unlike most -lactams carbenicillin disodium is limited to primarily gram-negative bacteria including Pseudomonas aeruginosa and common enteric species It can be used in selection protocols and is often preferred over ampicillin due to its increased stability This allows for less contamination of unwanted bacterial colonies In addition carbenicillin has been used to regulate bacterial growth in plants yielding low regeneration frequencies It can be used in genetic transformation applications to select for AmpR transformed cells