Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbenoxolone disodium | 7421-40-1 | 99.9% | 614.72 | C34H48Na2O7 | 10 MG
SDP

Catalog No. 5000298923
Encompass_Preferred

Carbenoxolone disodium is a purified research reagent used as a gap junction uncoupling agent to study intercellular communication, inflammation, and viral replication in vitro. It is supplied in small pack sizes for laboratory experiments and should be stored sealed and protected from moisture.

  • Used as a gap junction and hemichannel inhibitor in cell biology studies.
  • CAS number 7421-40-1 for unambiguous identification.
  • Molecular formula C34H48Na2O7 and molecular weight 614.72.
  • High purity (approximately 99.9%) suitable for research applications.
  • Storage: -20°C sealed; in solvent -80°C (1 year) or -20°C (6 months).
  • Available in small research pack sizes, including 10 mg.

Catalog No. 50-002-98923 Supplier Medchemexpress LLC Supplier No. HYB136710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Carbenoxolone disodium is a purified research reagent used as a gap junction uncoupling agent to study intercellular communication, inflammation, and viral replication in vitro. It is supplied in small pack sizes for laboratory experiments and should be stored sealed and protected from moisture.

  • Used as a gap junction and hemichannel inhibitor in cell biology studies.
  • CAS number 7421-40-1 for unambiguous identification.
  • Molecular formula C34H48Na2O7 and molecular weight 614.72.
  • High purity (approximately 99.9%) suitable for research applications.
  • Storage: -20°C sealed; in solvent -80°C (1 year) or -20°C (6 months).
  • Available in small research pack sizes, including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.