Learn More
Medchemexpress LLC Carbenoxolone disodium | 7421-40-1 | MFCD03840711 | 99.9% | 25 MG
SDP
Catalog No. 5000414100
Encompass_Preferred
Carbenoxolone disodium is the active metabolite of Glycyrrhizic acid and an inhibitor of human 11β-HSD and bacterial 3α, 20β-HSD. It functions as an uncoupling agent for gap junctions and is a potent inhibitor of Vaccinia virus replication. This product is used for research into peptic, esophageal, and oral ulceration and inflammation.
  • Active metabolite of Glycyrrhizic acid.
  • Inhibitor of human 11β-HSD and bacterial 3α, 20β-HSD.
  • Uncoupling agent for gap junctions.
  • Inhibitor of Vaccinia virus replication.
  • Used for studying peptic, esophageal, and oral ulceration and inflammation.
Catalog No. 50-004-14100 Supplier Medchemexpress LLC Supplier No. 25MGHYB1367
Add to Cart
Add to Cart
Carbenoxolone disodium is the active metabolite of Glycyrrhizic acid and an inhibitor of human 11β-HSD and bacterial 3α, 20β-HSD. It functions as an uncoupling agent for gap junctions and is a potent inhibitor of Vaccinia virus replication. This product is used for research into peptic, esophageal, and oral ulceration and inflammation.
  • Active metabolite of Glycyrrhizic acid.
  • Inhibitor of human 11β-HSD and bacterial 3α, 20β-HSD.
  • Uncoupling agent for gap junctions.
  • Inhibitor of Vaccinia virus replication.
  • Used for studying peptic, esophageal, and oral ulceration and inflammation.