Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Carbenoxolone Disodium Salt

Catalog No. 503119120 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-311-9120 1 g
50-311-9119 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 50-311-9120 Supplier Sigma Aldrich Supplier No. C47905G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Carbenoxolone disodium is a water soluble disodium salt of glycyrrhetinic acid hemisuccinate. It acts as an efficient promoter for the nasal absorption of insulin.

Specifications

CAS 17465-86-0
Molecular Formula C48H80O40
Linear Formula C48H80O40
MDL Number MFCD00009595
Quantity 1 g
Synonym 3beta-Hydroxy-11-oxoolean-12-en-30-oic acid 3-hemisuccinate
Molecular Weight (g/mol) 1297.12 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.