Learn More
Selleck Chemical LLC Carbenoxolone Sodium-25mg
SDP
Supplier:  Selleck Chemical LLC S436825MG
Encompass_Preferred
Carbenoxolone, a synthetic derivative of glycyrrhetinic acid, is also used as a blocker of the enzyme 11β-hydroxysteroid dehydrogenase (11β-HSD), of pannexon membrane channels (comprising 6 subunits of pannexin) and the related innexon channels.
Catalog No. 50-239-0036