Learn More
Cayman Chemical CarbetapentncItrate 100mg
SDP
Catalog No. 502965995
Encompass_Preferred
A σ1 receptor agonist (Ki = 10.4 nM); inhibits citric acid-induced cough in guinea pigs from 1-5 mg/kg; reduces the number of seizures, mortality, and hippocampal neuronal cell death in a rat model of kainate-induced seizures at 12.5 and 25 mg/kg
Catalog No. 50-296-5995 Supplier Cayman Chemical Supplier No. 18606100 MG
A σ1 receptor agonist (Ki = 10.4 nM); inhibits citric acid-induced cough in guinea pigs from 1-5 mg/kg; reduces the number of seizures, mortality, and hippocampal neuronal cell death in a rat model of kainate-induced seizures at 12.5 and 25 mg/kg