Learn More
CPC Scientific Butyryl-Tyr(Me)-Ile-Gln-Asn-Cys-Pro-Leu-Gly-NH2 (trifluoroacetate salt) 25MG
SDP
Catalog No. 502110047
Encompass
Carbetocin shows a prolonged effect due to an increased resistance to enzymatic cleavage. Carbetocin is highly effective in preventing post-partum hemorrhage after vaginal delivery.ONE-LETTER SEQUENCE: Butyryl-Y(Me)-INCPLG-NH2MOLECULAR FORMULA: C45H69N11O12S1MOLECULAR WEIGHT:988.18STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [37025-55-1], RESEARCH AREA: HormonalREFERENCES: T.Barth et al., Coll. Czech. Chem. Commun., 46, 2441 (1981); N.Cort et al., Am. J. Vet. Res., 43, 1283 (1982); V.Klenerova et al., J. Physiol. Pharmacol., 60, 57 (2009); N.C.Peters and J.J.Duvekot, Obstet. Gynecol. Surv., 64, 129 (2009); W.Rath, Eur. J. Obstet. Gynecol. Reprod. Biol., 147, 15 (2009)
Catalog No. 50-211-0047 Supplier CPC Scientific Supplier No. OXYT002B
Carbetocin shows a prolonged effect due to an increased resistance to enzymatic cleavage. Carbetocin is highly effective in preventing post-partum hemorrhage after vaginal delivery.ONE-LETTER SEQUENCE: Butyryl-Y(Me)-INCPLG-NH2MOLECULAR FORMULA: C45H69N11O12S1MOLECULAR WEIGHT:988.18STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [37025-55-1], RESEARCH AREA: HormonalREFERENCES: T.Barth et al., Coll. Czech. Chem. Commun., 46, 2441 (1981); N.Cort et al., Am. J. Vet. Res., 43, 1283 (1982); V.Klenerova et al., J. Physiol. Pharmacol., 60, 57 (2009); N.C.Peters and J.J.Duvekot, Obstet. Gynecol. Surv., 64, 129 (2009); W.Rath, Eur. J. Obstet. Gynecol. Reprod. Biol., 147, 15 (2009)