Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbetocin | 37025-55-1 | MFCD01076600 | 99.5% | 1048.21 | C47H73N11O14S | 10 MG
SDP

Catalog No. 5000299260
Encompass_Preferred

Carbetocin acetate is an oxytocin receptor agonist peptide supplied as a research reagent for preclinical studies, including postpartum hemorrhage and central oxytocin receptor activity. It is reported to cross the blood-brain barrier.

  • Research-grade oxytocin receptor agonist.
  • High purity (99.5%).
  • Molecular weight 1048.21 g/mol.
  • Chemical formula C47H73N11O14S.
  • White to off-white powder supplied as 10 mg.
  • Store sealed, protected from moisture and light; powder: -80°C for long term, -20°C for short term.

Catalog No. 50-002-99260 Supplier Medchemexpress LLC Supplier No. HY17573A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbetocin acetate is an oxytocin receptor agonist peptide supplied as a research reagent for preclinical studies, including postpartum hemorrhage and central oxytocin receptor activity. It is reported to cross the blood-brain barrier.

  • Research-grade oxytocin receptor agonist.
  • High purity (99.5%).
  • Molecular weight 1048.21 g/mol.
  • Chemical formula C47H73N11O14S.
  • White to off-white powder supplied as 10 mg.
  • Store sealed, protected from moisture and light; powder: -80°C for long term, -20°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.