Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Carbetocin acetate | 1631754-28-3 | 99.6% | 1048.21 | 100 MG
SDP

Catalog No. 5000387531
Encompass_Preferred

Carbetocin acetate is an oxytocin (OT) analogue and an oxytocin receptor agonist with a Ki of 7.1 nM. It has potential for postpartum hemorrhage research and can cross the blood-brain barrier, producing antidepressant-like activity via activation of oxytocin receptors in the CNS. In vitro, it demonstrates significantly higher stability and a longer duration of action compared to oxytocin.

  • Oxytocin analogue and receptor agonist
  • High affinity to oxytocin receptor (Ki=7.1 nM)
  • Potential for postpartum hemorrhage research
  • Crosses blood-brain barrier
  • Produces antidepressant-like activity
  • Higher stability and longer duration of action in vitro
  • Solid, white to off-white appearance
  • Chemical formula: C47H73N11O14S
  • CAS number: 1631754-28-3

Catalog No. 50-003-87531 Supplier Medchemexpress LLC Supplier No. HY17573A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbetocin acetate is an oxytocin (OT) analogue and an oxytocin receptor agonist with a Ki of 7.1 nM. It has potential for postpartum hemorrhage research and can cross the blood-brain barrier, producing antidepressant-like activity via activation of oxytocin receptors in the CNS. In vitro, it demonstrates significantly higher stability and a longer duration of action compared to oxytocin.

  • Oxytocin analogue and receptor agonist
  • High affinity to oxytocin receptor (Ki=7.1 nM)
  • Potential for postpartum hemorrhage research
  • Crosses blood-brain barrier
  • Produces antidepressant-like activity
  • Higher stability and longer duration of action in vitro
  • Solid, white to off-white appearance
  • Chemical formula: C47H73N11O14S
  • CAS number: 1631754-28-3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.