Learn More
Medchemexpress LLC Carbetocin (acetate) | 1631754-28-3 | 99.6% | 1048.21 | 5 MG
SDP
Catalog No. 5000386749
Encompass_Preferred
Carbetocin acetate is an oxytocin (OT) analogue and an oxytocin receptor agonist with a Ki of 7.1 nM. It shows high affinity to the chimeric N-terminus (E1) of the oxytocin receptor (Ki=1.17 μM). This compound has potential for research related to postpartum hemorrhage and can cross the blood-brain barrier, exhibiting antidepressant-like activity through the activation of oxytocin receptors in the central nervous system.
  • Oxytocin (OT) analogue and oxytocin receptor agonist.
  • Ki of 7.1 nM for oxytocin receptor.
  • High affinity to chimeric N-terminus (E1) of the oxytocin receptor (Ki=1.17 μM).
  • Potential for postpartum hemorrhage research.
  • Can cross the blood-brain barrier.
  • Produces antidepressant-like activity via activation of oxytocin receptors in the CNS.
  • Agonist with significantly higher stability and longer duration of action compared to oxytocin.
Catalog No. 50-003-86749 Supplier Medchemexpress LLC Supplier No. HY17573A5MG
Carbetocin acetate is an oxytocin (OT) analogue and an oxytocin receptor agonist with a Ki of 7.1 nM. It shows high affinity to the chimeric N-terminus (E1) of the oxytocin receptor (Ki=1.17 μM). This compound has potential for research related to postpartum hemorrhage and can cross the blood-brain barrier, exhibiting antidepressant-like activity through the activation of oxytocin receptors in the central nervous system.
  • Oxytocin (OT) analogue and oxytocin receptor agonist.
  • Ki of 7.1 nM for oxytocin receptor.
  • High affinity to chimeric N-terminus (E1) of the oxytocin receptor (Ki=1.17 μM).
  • Potential for postpartum hemorrhage research.
  • Can cross the blood-brain barrier.
  • Produces antidepressant-like activity via activation of oxytocin receptors in the CNS.
  • Agonist with significantly higher stability and longer duration of action compared to oxytocin.