Learn More
Medchemexpress LLC Carbetocin (acetate) | 1631754-28-3 | 99.6% | 1048.21 | 50 MG
SDP
Catalog No. 5000387138
Encompass_Preferred
Carbetocin acetate is an oxytocin (OT) analogue and an oxytocin receptor agonist with a Ki of 7.1 nM. It shows high affinity to the chimeric N-terminus (E1) of the oxytocin receptor (Ki=1.17 μM). This compound has potential for research related to postpartum hemorrhage. It can cross the blood-brain barrier and exhibits antidepressant-like activity by activating oxytocin receptors in the CNS.
  • Oxytocin receptor agonist
  • Oxytocin (OT) analogue
  • Potential for postpartum hemorrhage research
  • Can cross the blood-brain barrier
  • Produces antidepressant-like activity via activation of oxytocin receptors in the CNS
  • Higher stability and longer duration of action compared to oxytocin
Catalog No. 50-003-87138 Supplier Medchemexpress LLC Supplier No. HY17573A50MG
Carbetocin acetate is an oxytocin (OT) analogue and an oxytocin receptor agonist with a Ki of 7.1 nM. It shows high affinity to the chimeric N-terminus (E1) of the oxytocin receptor (Ki=1.17 μM). This compound has potential for research related to postpartum hemorrhage. It can cross the blood-brain barrier and exhibits antidepressant-like activity by activating oxytocin receptors in the CNS.
  • Oxytocin receptor agonist
  • Oxytocin (OT) analogue
  • Potential for postpartum hemorrhage research
  • Can cross the blood-brain barrier
  • Produces antidepressant-like activity via activation of oxytocin receptors in the CNS
  • Higher stability and longer duration of action compared to oxytocin