Learn More
Cayman Chemical Carbetocin (acetate), Purity: ≥98%, CAS: 1631754-28-3, 50 mg
SDP
Catalog No. 502946322
Encompass_Preferred
A peptide oxytocin receptor agonist; binds to oxytocin receptors (Kd = 1.96 nM); selectively binds to oxytocin receptors over vasopressin V2 receptors (Kds = 1.96 and 61.3 nM, respectively); induces contraction of isolated rat uterine strips (EC50 = 48 nM); inhibits oxytocin-induced contraction of isolated rat uterine strips (pA2 = 8.21); increases the frequency and amplitude of uterine contractions in early postpartum dairy cows at 0.35 mg/animal; prevents stress-induced reinstatement of morphine-seeking behavior in mice at 6.4 mg/kg; reverses learned helplessness and decreases social withdrawal induced by chronic unpredictable stress in tree shrews
Catalog No. 50-294-6322 Supplier Cayman Chemical Supplier No. 3323450 MG
A peptide oxytocin receptor agonist; binds to oxytocin receptors (Kd = 1.96 nM); selectively binds to oxytocin receptors over vasopressin V2 receptors (Kds = 1.96 and 61.3 nM, respectively); induces contraction of isolated rat uterine strips (EC50 = 48 nM); inhibits oxytocin-induced contraction of isolated rat uterine strips (pA2 = 8.21); increases the frequency and amplitude of uterine contractions in early postpartum dairy cows at 0.35 mg/animal; prevents stress-induced reinstatement of morphine-seeking behavior in mice at 6.4 mg/kg; reverses learned helplessness and decreases social withdrawal induced by chronic unpredictable stress in tree shrews