Learn More
Electron Microscopy Sciences Optical Fiber Carbide Cleaver Tool
SDP
Catalog No. 5031523
Encompass_Preferred
Made from a solid carbide rod with two angled sides which intersect in a very sharp cutting edge of about 4mm in length. Shank is mounted into a non-rolling hexagon shaped anodized aluminum holder. Carbide is 2.0mm dia x 10.0mm.
Catalog No. 50-315-23 Supplier Electron Microscopy Sciences Supplier No. 62165CC
Made from a solid carbide rod with two angled sides which intersect in a very sharp cutting edge of about 4mm in length. Shank is mounted into a non-rolling hexagon shaped anodized aluminum holder. Carbide is 2.0mm dia x 10.0mm.