Learn More
Med Vet International Carbide Float Blade, Coarse, J0057C, Each
Catalog No. 5000584019
Encompass_Preferred
These blades take alot of the work out of floating a horse’s teeth. The carbide metal is very hard and gives a razor sharp edge. This is the new standard in equine dentistry. Blade can be resharpened 3 times. Available in 3 different grits: fine, medium, and coarse. Medium is the most common grit used. Fits any standard flat headed float. 3 1/4" x 1".
Catalog No. 50-005-84019 Supplier Med Vet International Supplier No. JOR57C
These blades take alot of the work out of floating a horse’s teeth. The carbide metal is very hard and gives a razor sharp edge. This is the new standard in equine dentistry. Blade can be resharpened 3 times. Available in 3 different grits: fine, medium, and coarse. Medium is the most common grit used. Fits any standard flat headed float. 3 1/4" x 1".