Learn More
Electron Microscopy Sciences Carbide Micro Tool Set 0.50 mm
SDP
Catalog No. 50996702
Encompass
These unique micro cutting tools are made using the latest grinding and polishing technology producing the highest quality tool on the market today. Each set comes with a MT-1 Handle, and instrument case. Tips are Micro-Needle, Micro-Graver, 3-sided, Micro-Chisel, Micro-Spade, Micro-Knife 20 degree , Micro-Knife 45 degree ,Micro-Graver Oval, and Micro-Scraper
Tip: 0.5 mm (.020")
Catalog No. 50-996-702 Supplier Electron Microscopy Sciences Supplier No. 6209550
These unique micro cutting tools are made using the latest grinding and polishing technology producing the highest quality tool on the market today. Each set comes with a MT-1 Handle, and instrument case. Tips are Micro-Needle, Micro-Graver, 3-sided, Micro-Chisel, Micro-Spade, Micro-Knife 20 degree , Micro-Knife 45 degree ,Micro-Graver Oval, and Micro-Scraper
Tip: 0.5 mm (.020")