Learn More
Surgical Design Glass Cutter with T.C. Blade Wheel
Catalog No. 11326 Shop All Surgical Design Products
Change view
Click to view available options
For Use With (Application):
Designed for cutting borosilicate glass easily, breaking off single- or double-strength glass
Catalog No. For Use With (Application)
11-326 Designed for cutting borosilicate glass easily, breaking off single- or double-strength glass
1 options
This item is not returnable. View return policy
Catalog No. 11-326 Supplier Surgical Design Supplier No. 33226
Add to Cart
Add to Cart
This item is not returnable. View return policy
Glass cutter designed for long lasting use.
  • Glass Cutter with Tungsten Carbide Wheel is made from Tungsten Carbide (T.C.) which prevents compromise of the blade wheel for long lasting use!
  • Glass Cutter Makes An Excellent Score Every Time
  • Special Tungsten-Carbide (T.C.) inserts for long-lasting use.
Certifications
ASTM, ISO 9001, cGMP
Specifications
For Use With (Application) Designed for cutting borosilicate glass easily, breaking off single- or double-strength glass
Product Type Glass Cutter