Learn More
Apexbio Technology LLC Carbidopa 28860-95-9 25mg
SDP
Catalog No. 502240396
Encompass_Preferred
Carbidopa (CAS 28860-95-9) is a peripheral aromatic L-amino acid decarboxylase inhibitor that limits the peripheral conversion of levodopa to dopamine thereby increasing the availability of levodopa for transport into the central nervous system In addition to its established role in Parkinson s disease mechanism studies Carbidopa has been identified as a selective modulator of the aryl hydrocarbon receptor (AhR) influencing downstream signaling pathways Recent studies indicate its potential to suppress the proliferation of pancreatic cancer cells and inhibit tumor growth supporting its use in investigations related to tumor biology and anticancer pharmacology
This item is not returnable. View return policy
Catalog No. 50-224-0396 Supplier Apexbio Technology LLC Supplier No. B190425
Carbidopa (CAS 28860-95-9) is a peripheral aromatic L-amino acid decarboxylase inhibitor that limits the peripheral conversion of levodopa to dopamine thereby increasing the availability of levodopa for transport into the central nervous system In addition to its established role in Parkinson s disease mechanism studies Carbidopa has been identified as a selective modulator of the aryl hydrocarbon receptor (AhR) influencing downstream signaling pathways Recent studies indicate its potential to suppress the proliferation of pancreatic cancer cells and inhibit tumor growth supporting its use in investigations related to tumor biology and anticancer pharmacology