Learn More
TARGETMOL CHEMICALS INC CARBIDOPA 50MG
SDP
Catalog No. 502576453
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carbidopa (Lodosyn) is an aromatic-L-amino-acid decarboxylase inhibitor (IC50 29±2 uM). It is used in PARKINSON DISEASE to reduce peripheral adverse effects of LEVODOPA. purity: 98%
Catalog No. 50-257-6453 Supplier TARGETMOL CHEMICALS INC Supplier No. T679550MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Carbidopa (Lodosyn) is an aromatic-L-amino-acid decarboxylase inhibitor (IC50 29±2 uM). It is used in PARKINSON DISEASE to reduce peripheral adverse effects of LEVODOPA. purity: 98%