Learn More
Medchemexpress LLC Carbidopa-d3 monohydrate | 1276197-58-0 | 98.5% | 1 MG
SDP
Catalog No. 5000406732
Encompass_Preferred
Carbidopa-d3 (monohydrate) is the deuterium labeled Carbidopa monohydrate. Carbidopa ((S)-(-)-Carbidopa) monohydrate, a peripheral decarboxylase inhibitor, can be used for the research of Parkinson's disease. Additionally, Carbidopa monohydrate is a selective aryl hydrocarbon receptor (AhR) modulator and has shown to inhibit pancreatic cancer cell and tumor growth.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06732 Supplier Medchemexpress LLC Supplier No. HYB0311AS1MG
Carbidopa-d3 (monohydrate) is the deuterium labeled Carbidopa monohydrate. Carbidopa ((S)-(-)-Carbidopa) monohydrate, a peripheral decarboxylase inhibitor, can be used for the research of Parkinson's disease. Additionally, Carbidopa monohydrate is a selective aryl hydrocarbon receptor (AhR) modulator and has shown to inhibit pancreatic cancer cell and tumor growth.
  • Used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Deuteration has the potential to influence the pharmacokinetic and metabolic profiles of drugs.