Learn More
Cayman Chemical Carbidopa-d3 (hydrate), Purity: ≥99% deuterated forms (d1-d3), CAS: 1276197-58-0, 5 mg
SDP
Catalog No. 502910330
Encompass_Preferred
An internal standard for the quantification of carbidopa by GC- or LC-MS
Catalog No. 50-291-0330 Supplier Cayman Chemical Supplier No. 348845 MG
An internal standard for the quantification of carbidopa by GC- or LC-MS