Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbidopa Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014103 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4103 Supplier MilliporeSigma Supelco Supplier No. PHR16551G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carbidopa is a peripheral decarboxylase inhibitor which is often formulated with levodopa for the management of the symptoms of Parkinson′s disease. Its mode of action involves the reduction of systemic production of dopamine and minimization of undesirable effects such as nausea.Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 38821-49-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H14N2O4 · H2O
MDL Number MFCD00889211
Synonym (S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid monohydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 244.24
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.