Learn More
Cayman Chemical CarbImazole 100g
SDP
Catalog No. 502941155
Encompass_Preferred
A prodrug form of methimazole; reduces total thyroxine concentration, thyroid enlargement, polyphagia, anorexia, and alopecia in a cat model of hyperthyroidism at 15 mg/animal
Catalog No. 50-294-1155 Supplier Cayman Chemical Supplier No. 28280100 G
A prodrug form of methimazole; reduces total thyroxine concentration, thyroid enlargement, polyphagia, anorexia, and alopecia in a cat model of hyperthyroidism at 15 mg/animal