Learn More
Medchemexpress LLC Carbimazole | 22232-54-8 | 98.0% | 1 ML
SDP
Catalog No. 5000407901
Encompass_Preferred
Carbimazole is an orally active antithyroid agent that converts to Methimazole after absorption. It reduces the production of thyroxine by preventing the thyroid peroxidase enzyme from iodinating and coupling tyrosine residues on thyroglobulin. This agent also exhibits anti-inflammatory and neuronal-protective activities.
  • Orally active antithyroid agent.
  • Converts to Methimazole in the body.
  • Reduces thyroxine production.
  • Displays anti-inflammatory activity.
  • Shows neuronal-protective activity.
  • Suggested for application in hyperthyroidism and neurological research.
Catalog No. 50-004-07901 Supplier Medchemexpress LLC Supplier No. HYB055810MM/1ML
Carbimazole is an orally active antithyroid agent that converts to Methimazole after absorption. It reduces the production of thyroxine by preventing the thyroid peroxidase enzyme from iodinating and coupling tyrosine residues on thyroglobulin. This agent also exhibits anti-inflammatory and neuronal-protective activities.
  • Orally active antithyroid agent.
  • Converts to Methimazole in the body.
  • Reduces thyroxine production.
  • Displays anti-inflammatory activity.
  • Shows neuronal-protective activity.
  • Suggested for application in hyperthyroidism and neurological research.