Learn More
Selleck Chemical LLC CarbinoxaMine Maleate S4696-100mg
SDP
Catalog No. 503109166
Encompass_Preferred
Carbinoxamine maleate is a histamine-H1 receptor blocking agent with antihistamine and anticholinergic properties
Catalog No. 50-310-9166 Supplier Selleck Chemical LLC Supplier No. S4696100MG
Carbinoxamine maleate is a histamine-H1 receptor blocking agent with antihistamine and anticholinergic properties