Learn More
Cayman Chemical Carbmylatd Alph1 ntItryp 500uL
SDP
Catalog No. 502933107
Encompass_Preferred
An inhibitor of ATG4B inhibits ATG4B cleavage of LC3B-GST (IC50 51 UM) and reduces LC3B lipidation induced by starvation in Saos-2 cells reduces starvation-induced autophagy and prevents autophagic vacuole formation in Saos-2 and MDA-MB-468 cells (100 UM) decreases autophagy in vivo in mouse liver inhibits tumor growth in mouse xenograft model and decreases the number of autophagic vacuoles in osteosarcoma tumors (100 mg/kg)
Catalog No. 50-293-3107 Supplier Cayman Chemical Supplier No. 25340500 UL
An inhibitor of ATG4B inhibits ATG4B cleavage of LC3B-GST (IC50 51 UM) and reduces LC3B lipidation induced by starvation in Saos-2 cells reduces starvation-induced autophagy and prevents autophagic vacuole formation in Saos-2 and MDA-MB-468 cells (100 UM) decreases autophagy in vivo in mouse liver inhibits tumor growth in mouse xenograft model and decreases the number of autophagic vacuoles in osteosarcoma tumors (100 mg/kg)