Learn More
Thermo Scientific CARBO PK MA1G GUARD COLMN 1/EA
Catalog No. NC9459597 Shop All Thermo Scientific Products
Encompass
CarboPac MA1 Guard Column (4 x 50 mm)
Catalog No. NC9459597 Supplier Thermo Scientific Supplier No. 044067
CarboPac MA1 Guard Column (4 x 50 mm)