Learn More
Medchemexpress LLC Carbobenzoxy-L-leucyl-L-leucyl-L-glutamic acid 7-amido-4-methylcoumarin | 348086-66-8 | MFCD01074989 | 99.5% | 664.75 | C35H44N4O9 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1230535MG
Encompass_Preferred
Z-Leu-Leu-Glu-AMC is a fluorogenic tripeptide substrate used to measure caspase-like (peptidylglutamyl-peptide hydrolyzing) activity of the 20S proteasome. Proteolytic cleavage releases 7-amino-4-methylcoumarin (AMC), generating a fluorescent signal for biochemical proteasome activity assays and inhibitor screening.
  • Fluorogenic substrate releasing AMC for fluorescence detection.
  • Suitable for measuring caspase-like/PGPH activity of the 20S proteasome.
  • High purity appropriate for biochemical assays and screening.
  • Available in multiple sizes and solution formats for assay flexibility.
  • Compatible with standard assay buffers and fluorescence detection setups.
Catalog No. 50-002-38710