Learn More
Selleck Chemical LLC Carbocisteine S5216-25mg
SDP
Catalog No. 503112204
Encompass_Preferred
Carbocisteine (S-carboxymethyl-l-cysteine) is a very important pharmaceutical product with mucolitic activity and is normally used for the treatment of bronchitis and colds
Catalog No. 50-311-2204 Supplier Selleck Chemical LLC Supplier No. S521625MG
Carbocisteine (S-carboxymethyl-l-cysteine) is a very important pharmaceutical product with mucolitic activity and is normally used for the treatment of bronchitis and colds