Learn More
Medchemexpress LLC Carbocysteine sulfoxide | 5439-87-2 | 195.19 | C5H9NO5S | 5 MG
SDP
Catalog No. 5000281181
Encompass_Preferred
Carbocysteine sulfoxide is a carbocisteine impurity and drug intermediate provided for research use only. The compound has formula C5H9NO5S, molecular weight 195.19, and CAS number 5439-87-2. It is intended for analytical reference, impurity profiling, and synthetic studies in laboratory research and is not for clinical, diagnostic, or therapeutic use.
  • Used as an analytical reference material for method validation.
  • Useful for impurity profiling and degradation studies.
  • Suitable as a synthetic intermediate in research workflows.
  • Provided in small, accurately measured quantities for laboratory experiments.
  • Intended strictly for research use; not for human or animal administration.
Catalog No. 50-002-81181 Supplier Medchemexpress LLC Supplier No. HYW4223625MG
Carbocysteine sulfoxide is a carbocisteine impurity and drug intermediate provided for research use only. The compound has formula C5H9NO5S, molecular weight 195.19, and CAS number 5439-87-2. It is intended for analytical reference, impurity profiling, and synthetic studies in laboratory research and is not for clinical, diagnostic, or therapeutic use.
  • Used as an analytical reference material for method validation.
  • Useful for impurity profiling and degradation studies.
  • Suitable as a synthetic intermediate in research workflows.
  • Provided in small, accurately measured quantities for laboratory experiments.
  • Intended strictly for research use; not for human or animal administration.