Learn More
Cayman Chemical Carbofurn-d3 500ug
SDP
Catalog No. 502919237
Encompass_Preferred
An internal standard for the quantification of carbofuran by GC-or LC-MS
Catalog No. 50-291-9237 Supplier Cayman Chemical Supplier No. 39558500 UG
An internal standard for the quantification of carbofuran by GC-or LC-MS