Learn More
Cusabio Technology LLC carbohydrate antigen 5
Catalog No. 5000051510
Encompass_Preferred
This kit detects CA50 in human serum plasma cell culture supernates ascitic fluid tissue homogenates and cell lysates using a sandwich ELISA method It offers a quantitative detection range of 10 U/mL to 120 U/mL with a sensitivity of 2 5 U/mL ensuring precise measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it efficient for routine laboratory workflows Absorbance is measured at 450 nm for accurate results Designed for research and diagnostic applications this kit provides reliable performance for analyzing CA50 levels in diverse biological matrices supporting studies related to cancer biomarkers and disease monitoring
Catalog No. 50-000-51510 Supplier Cusabio Technology LLC Supplier No. CSBEQ027409HU
This kit detects CA50 in human serum plasma cell culture supernates ascitic fluid tissue homogenates and cell lysates using a sandwich ELISA method It offers a quantitative detection range of 10 U/mL to 120 U/mL with a sensitivity of 2 5 U/mL ensuring precise measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it efficient for routine laboratory workflows Absorbance is measured at 450 nm for accurate results Designed for research and diagnostic applications this kit provides reliable performance for analyzing CA50 levels in diverse biological matrices supporting studies related to cancer biomarkers and disease monitoring