Learn More
Waters Corp Carbohydrate Cartridge Column, 4.6 mm, 250 mm, 1/pk, with eConnect Column Tag
Catalog No. 502392080 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This carbohydrate cartridge column, packed with 4-μm spherical silica, is designed for the high-speed separation of five monosaccharides and disaccharides. It features an electronic eConnect column tag for automated tracking and is compatible with any HPLC system.
  • Packed with 4-μm spherical silica.
  • Separates five monosaccharides and disaccharides with baseline resolution in under 12 minutes.
  • Pre-packed, disposable cartridge column.
  • Requires reusable end-fittings (available separately).
  • Electronic eConnect column tag provides unique, specific information.
  • NFC-enabled tags automatically identify, track, and trace column use history.
  • Usable on any HPLC.
  • Advantages unlocked with compatible systems running Empower software.
Catalog No. 50-239-2080 Supplier Waters Corp Supplier No. WAT044355RF
Add to Cart
Add to Cart
This carbohydrate cartridge column, packed with 4-μm spherical silica, is designed for the high-speed separation of five monosaccharides and disaccharides. It features an electronic eConnect column tag for automated tracking and is compatible with any HPLC system.
  • Packed with 4-μm spherical silica.
  • Separates five monosaccharides and disaccharides with baseline resolution in under 12 minutes.
  • Pre-packed, disposable cartridge column.
  • Requires reusable end-fittings (available separately).
  • Electronic eConnect column tag provides unique, specific information.
  • NFC-enabled tags automatically identify, track, and trace column use history.
  • Usable on any HPLC.
  • Advantages unlocked with compatible systems running Empower software.