Learn More
Waters Corp Waters Carbohydrate Cartridge Column, 4.6 mm ID × 250 mm, 4 μm
Catalog No. 50785568 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This High-Performance Carbohydrate Cartridge Column is packed with 4-μm spherical silica. It was developed for baseline resolution separation of five monosaccharides and disaccharides in under 12 minutes. The pre-packed, disposable cartridge column requires reusable end-fittings, sold separately. It aids in controlling food and beverage quality and ensuring regulatory compliance.
  • Separates five monosaccharides and disaccharides with baseline resolution in under 12 minutes.
  • Features 4-μm spherical silica packing.
  • Pre-packed, disposable cartridge format.
  • Requires reusable end-fittings (sold separately).
  • Offers optimal speed, resolution, and longevity.
  • Reversed-phase mode for separating monosaccharides and oligosaccharides up to DP 8.
  • Supports analysis and quantification of food and beverage ingredients.
Catalog No. 50-785-568 Supplier Waters Corp Supplier No. WAT044355
This High-Performance Carbohydrate Cartridge Column is packed with 4-μm spherical silica. It was developed for baseline resolution separation of five monosaccharides and disaccharides in under 12 minutes. The pre-packed, disposable cartridge column requires reusable end-fittings, sold separately. It aids in controlling food and beverage quality and ensuring regulatory compliance.
  • Separates five monosaccharides and disaccharides with baseline resolution in under 12 minutes.
  • Features 4-μm spherical silica packing.
  • Pre-packed, disposable cartridge format.
  • Requires reusable end-fittings (sold separately).
  • Offers optimal speed, resolution, and longevity.
  • Reversed-phase mode for separating monosaccharides and oligosaccharides up to DP 8.
  • Supports analysis and quantification of food and beverage ingredients.