Learn More
Description
- A normal test diet modified to contain 68% carbohydrate by weight
Specifications
Specifications
| Product Type | Carbohydrate Diet |
| Format | Pelleted |
| Packaging Quantity | 10 kg |
| Storage Requirements | Store at 2°C to 8°C |
Safety and Handling
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.