Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Carbohydrazide 25 GR
SDP

Catalog No. 50980287
Encompass_Preferred

(Dimethylarsinic Acid)
CAS #497-18-7 CO(NHNH₂)₂ F.W. 90.08
Melting point: 152-153°C

For GACH embedding kit. A water-miscible, lipid-retaining, embedding polymer for E.M.. (Heckman, et. al., Ultrastruct. Res., 42,156, 1973).

Catalog No. 50-980-287 Supplier Electron Microscopy Sciences Supplier No. 12350
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Dimethylarsinic Acid)
CAS #497-18-7 CO(NHNH₂)₂ F.W. 90.08
Melting point: 152-153°C

For GACH embedding kit. A water-miscible, lipid-retaining, embedding polymer for E.M.. (Heckman, et. al., Ultrastruct. Res., 42,156, 1973).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.