Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbohydrazide, 97%

Catalog No. AC403975000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403975000 500 g Glass bottle
AC403970250 25 g Glass bottle
AC403971000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC403975000 Supplier Thermo Scientific Chemicals Supplier No. 403975000
Add to Cart

CAS: 497-18-7 | CH6N4O | 90.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-18-7
Molecular Formula CH6N4O
Molecular Weight (g/mol) 90.09
MDL Number MFCD00007591
InChI Key XEVRDFDBXJMZFG-UHFFFAOYSA-N
Synonym carbohydrazide, carbonic dihydrazide, carbazide, carbonohydrazide, carbodihydrazide, hydrazine, carbonyldi, hydrazine, carbonylbis, carbazic acid, hydrazide, semicarbazide, 4-amino, carbonic acid, dihydrazide
PubChem CID 73948
ChEBI CHEBI:61308
IUPAC Name 1,3-diaminourea
SMILES NNC(=O)NN

Specifications

Melting Point >153.0°C
Color White
Density 1.0200g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula NH2NHCONHNH2
Quantity 500 g
Beilstein 03,121
Specific Gravity 1.02
Merck Index 15,1804
Formula Weight 90.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Carbohydrazide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-837-2
RTECSNumber FF2625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.