Learn More
Newcomer Supply Carbol Fuchsin Stain, Kinyoun, 250 ml
SDP
Catalog No. NC1751763
Encompass
Newcomer Supply Carbol Fuchsin Stain, Kinyoun is used in the Kinyoun AFB Stain to demonstrate the presence of acid-fast mycobacteria in tissue sections. Carbol Fuchsin Stain, Kinyoun is a concentrated phenol and basic fuchsin solution that works to permeate the lipoid capsule of acid-fast organisms, rendering them resistant to acid alcohol decolorization. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC1751763 Supplier Newcomer Supply Supplier No. 1031A
Newcomer Supply Carbol Fuchsin Stain, Kinyoun is used in the Kinyoun AFB Stain to demonstrate the presence of acid-fast mycobacteria in tissue sections. Carbol Fuchsin Stain, Kinyoun is a concentrated phenol and basic fuchsin solution that works to permeate the lipoid capsule of acid-fast organisms, rendering them resistant to acid alcohol decolorization. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.