Learn More
Medchemexpress LLC Carbomer 934 | 9007-20-9 | MFCD00084394 | 95.0% | (C3H4O2)n | 25 G
SDP
Catalog No. 5000281521
Encompass_Preferred
Carbomer 934 is a crosslinked poly(acrylic acid) polymer used as a thickening, suspending, and gel-forming agent in pharmaceutical and topical formulations. It forms high-viscosity gels and provides a stable gel matrix with good resistance to heat, light, and microbial contamination.
  • Forms high-viscosity gels.
  • Provides a stable gel matrix for formulations.
  • Good heat, light, and microbial resistance.
  • Suitable as thickening and suspending agent for topical and pharmaceutical use.
  • White to off-white powder, typically supplied in 25 G packaging.
  • Purity ≥95.0% as stated on batch certificate of analysis.
Catalog No. 50-002-81521 Supplier Medchemexpress LLC Supplier No. HYW250721B25G
Carbomer 934 is a crosslinked poly(acrylic acid) polymer used as a thickening, suspending, and gel-forming agent in pharmaceutical and topical formulations. It forms high-viscosity gels and provides a stable gel matrix with good resistance to heat, light, and microbial contamination.
  • Forms high-viscosity gels.
  • Provides a stable gel matrix for formulations.
  • Good heat, light, and microbial resistance.
  • Suitable as thickening and suspending agent for topical and pharmaceutical use.
  • White to off-white powder, typically supplied in 25 G packaging.
  • Purity ≥95.0% as stated on batch certificate of analysis.