Learn More
Medchemexpress LLC Carbomer 934 (CBM 934) | 9007-20-9 | 95.0% | Nominally 3,000,000 | Poly(acrylic acid), repeating unit (C3H4O2)n | 100 G
SDP
Catalog No. 5000281644
Encompass_Preferred
Carbomer 934 (CBM 934) is a cross-linked poly(acrylic acid) polymer supplied as a powder for formulation use. It functions as a high-efficiency thickening and gelling agent, forming stable, high-viscosity gels suitable for topical, oral, and controlled-release pharmaceutical formulations. The material disperses in aqueous systems and can be neutralized to adjust viscosity and gel properties.
  • High thickening efficiency at low concentrations.
  • Forms stable, high-viscosity gels.
  • Suitable for controlled-release and topical formulations.
  • Dispersible in aqueous systems for easy processing.
  • Resistant to heat, light, and microbial contamination.
Catalog No. 50-002-81644 Supplier Medchemexpress LLC Supplier No. HYW250721B100G
Carbomer 934 (CBM 934) is a cross-linked poly(acrylic acid) polymer supplied as a powder for formulation use. It functions as a high-efficiency thickening and gelling agent, forming stable, high-viscosity gels suitable for topical, oral, and controlled-release pharmaceutical formulations. The material disperses in aqueous systems and can be neutralized to adjust viscosity and gel properties.
  • High thickening efficiency at low concentrations.
  • Forms stable, high-viscosity gels.
  • Suitable for controlled-release and topical formulations.
  • Dispersible in aqueous systems for easy processing.
  • Resistant to heat, light, and microbial contamination.