Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbomer 934P, Resin, NF, Spectrum™ Chemical
SDP

Catalog No. 18601453 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
22 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-453 22 kg Poly Bottle
18-601-450 100 g Poly Bottle
18-601-451 500 g Poly Bottle
18-601-452 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-453 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CA18422KGBL

missing translation for 'validMainframeMsg'

CA184, 9003-01-4

Spectrum™ Chemical Carbomer 934P, Resin, NF polymers can provide bioadhesion, taste-masking and good binding characteristics that provide highly effective controlled-release properties. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-01-4

Specifications

CAS Max % 100%
Quantity 22 kg
Loss on Drying 2%
Packaging Poly Bottle
Grade NF
Chemical Name or Material Carbomer 934P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.