Learn More
Medchemexpress LLC Carbomer 940 | 9007-20-9 | 95.0% | 4,000,000 g/mol | (C3H4O2)n | 250 MG
SDP
Catalog No. 5000280625
Encompass_Preferred
Carbomer 940 is a high-molecular-weight, cross-linked polyacrylic acid polymer used as a thickening agent and gelling excipient for laboratory and formulation research. It forms a stable, high-viscosity gel matrix suitable for pharmaceutical and cosmetic formulations.
  • Forms a high-viscosity gel matrix.
  • Provides good resistance to heat, light, and microbial contamination.
  • Acts as a thickening agent and gelling excipient for topical and formulation use.
  • Supplied as a white powder in small research pack sizes, including 250 mg.
  • Typical assay 95.0%.
  • Consult the safety data sheet for handling, storage, and impurity information.
Catalog No. 50-002-80625 Supplier Medchemexpress LLC Supplier No. HYW250721A250MG
Carbomer 940 is a high-molecular-weight, cross-linked polyacrylic acid polymer used as a thickening agent and gelling excipient for laboratory and formulation research. It forms a stable, high-viscosity gel matrix suitable for pharmaceutical and cosmetic formulations.
  • Forms a high-viscosity gel matrix.
  • Provides good resistance to heat, light, and microbial contamination.
  • Acts as a thickening agent and gelling excipient for topical and formulation use.
  • Supplied as a white powder in small research pack sizes, including 250 mg.
  • Typical assay 95.0%.
  • Consult the safety data sheet for handling, storage, and impurity information.