Learn More
Medchemexpress LLC Carbomer 980 | 9007-20-9 | MFCD00146299 | 280.74668 g·mol⁻¹ | (C3H4O2)n | 25 G
SDP
Catalog No. 5000281211
Encompass_Preferred
Carbomer 980 is a cross-linked poly(acrylic acid) polymer used as a thickening and gelling agent for laboratory and formulation applications. Supplied as a white powder, it is intended for preparing stable gel matrices and adjusting viscosity in aqueous and hydroalcoholic systems.
  • Effective thickener at low concentration
  • Forms clear, stable gels and viscous matrices
  • Compatible with aqueous and hydroalcoholic formulations
  • Powder form for accurate dosing and easy dispersion
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years)
Catalog No. 50-002-81211 Supplier Medchemexpress LLC Supplier No. HYW25072125G
Carbomer 980 is a cross-linked poly(acrylic acid) polymer used as a thickening and gelling agent for laboratory and formulation applications. Supplied as a white powder, it is intended for preparing stable gel matrices and adjusting viscosity in aqueous and hydroalcoholic systems.
  • Effective thickener at low concentration
  • Forms clear, stable gels and viscous matrices
  • Compatible with aqueous and hydroalcoholic formulations
  • Powder form for accurate dosing and easy dispersion
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years)